Learn More
3M™ Peltor™ Hygiene Kit for Earmuffs X Series
Brand: 3M™ 7000104049
Additional Details : Poids : 0.35000kg
Description
The 3M™ Peltor™ X-Series HYX1 Hygiene Kit includes replaceable ear cushions and foam liners for X-Series X1 Earmuffs, models X1A and X1P3E (sold separately)- Lightweight, low-profile earmuff designed for protection against low-level industrial noise and other loud sounds with a Noise Reduction Rating (NRR) of 22 dB. Class A
- Service kit will help extend the service life of hearing protection equipment and potentially result in cost savings
- Exposure to loud noises can adversely affect the ability to hear over time
- Wearing proper hearing protection devices can help in reducing the harmful effects of noise
- Sound levels are expressed in decibels, often abbreviated as dB
- Higher decibel numbers indicate louder sounds
- Typical decibel levels associated with common sounds include: outdoors with no people or traffic 20 dB, normal conversation 60 dB, power lawn mower 90 dB, chain saw 110 dB, and 12-gauge shotgun 165 dB
- Hearing protector NRR can be used to evaluate different hearing protection options
- Actual level of protection varies depending on fit, wearing time and other factors
- Fits a wide range of industrial safety hard hats, including 3M™ H-700 Series Hard Hats (sold separately)
- Low-profile cup design for improved compatibility with other personal protection equipment (PPE)
- Twin headband design helps reduce heat buildup with good fit and balance
- Attaches directly to many hard hats without an adapter
- Earcups tilt to ensure a customized fit and comfort for individual users
- 3M Catalog# HYX1, HYX2, HYX3, HYX4, HYX5
Specifications
For Peltor Earmuffs X5 | |
Replaceable ear cushions and foam liners | |
HYX5 | |
Ear Muff Hygiene Kit |