Learn More
3M™ Versaflo™ TR-600 Series PAPR Accessory, Filter Latch Assembly
For use with Versaflo™ TR-600 Series PAPR.
Brand: 3M™ 7100134950
14.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.90000kg
Description
- TR-651 filter launch assembly is replacement part for TR-600 motor/blower TR-602N
- Enables simple maintenance of TR-600
- Works with all Versaflo™ TR-6000 series filters and cartridges
- Blue
- 3M Catalog# TR-651
Applications:
Healthcare, Pharmaceutical
Certifications
NIOSH Approved
Specifications
Blue | |
Latch Assembly | |
Versaflo™ TR-600 Series PAPR System | |
1 Ea. |
Reusable | |
Healthcare, Pharmaceutical | |
TR-651 |