missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%, Thermo Scientific Chemicals

Catalog Number p-3777438
55.20 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
163439-82-5
MFCD01073894
QJRIUWQPJVPYSO-GHMZBOCLSA-N
3r,4r-1-benzylpyrrolidine-3,4-diol, 3r,4r---1-benzyl-3,4-pyrrolidindiol, 3r,4r---1-benzyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r, pubchem14982, trans-3,4-dihydroxy-n-benzylpyrrolidine, 3r,4r-1-benzyl-3,4-pyrrolidinediol, 3r-1-benzylpyrrolidine-3beta,4alpha-diol, 3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r
7021491
(3R,4R)-1-benzylpyrrolidine-3,4-diol
C1C(C(CN1CC2=CC=CC=C2)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
163439-82-5
MFCD01073894
QJRIUWQPJVPYSO-GHMZBOCLSA-N
3r,4r-1-benzylpyrrolidine-3,4-diol, 3r,4r---1-benzyl-3,4-pyrrolidindiol, 3r,4r---1-benzyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r, pubchem14982, trans-3,4-dihydroxy-n-benzylpyrrolidine, 3r,4r-1-benzyl-3,4-pyrrolidinediol, 3r-1-benzylpyrrolidine-3beta,4alpha-diol, 3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r
7021491
(3R,4R)-1-benzylpyrrolidine-3,4-diol
C1C(C(CN1CC2=CC=CC=C2)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 101.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H15NO2
Quantity 1 g
Specific Rotation − 32.00
Molecular Weight (g/mol) 193.24
Formula Weight 193.24
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material (3R, 4R)-(-)-1-Benzyl-3, 4-pyrrolidindiol
Show More Show Less
compliance-icons
Product Identifier
  • (3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'