missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3541124
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
179055-27-7
C11H10N2O
186.21
MFCD08669906
QWKUTEQPAJGRLZ-UHFFFAOYSA-N
4-1-methyl-1h-pyrazol-3-yl benzaldehyde, 4-1-methylpyrazol-3-yl benzaldehyde, benzaldehyde,4-1-methyl-1h-pyrazol-3-yl, benzaldehyde, 4-1-methyl-1h-pyrazol-3-yl, 3-4-formylphenyl-1-methyl-1h-pyrazole, 4-l-methyl-1h-pyrazol-3-yl benzaldehyde
10535545
CN1C=CC(=N1)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
179055-27-7
C11H10N2O
186.21
MFCD08669906
QWKUTEQPAJGRLZ-UHFFFAOYSA-N
4-1-methyl-1h-pyrazol-3-yl benzaldehyde, 4-1-methylpyrazol-3-yl benzaldehyde, benzaldehyde,4-1-methyl-1h-pyrazol-3-yl, benzaldehyde, 4-1-methyl-1h-pyrazol-3-yl, 3-4-formylphenyl-1-methyl-1h-pyrazole, 4-l-methyl-1h-pyrazol-3-yl benzaldehyde
10535545
CN1C=CC(=N1)C1=CC=C(C=O)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-(1-methyl-1H-pyrazol-3-yl)benzaldehyde
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'