missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7035926
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
82925-02-8
C19H24N2O2
312.41
MFCD04974539
DGOOLMGPMIHRFY-UHFFFAOYSA-N
4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamine
10041227
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1
This item is not returnable. View return policy
82925-02-8
C19H24N2O2
312.41
MFCD04974539
DGOOLMGPMIHRFY-UHFFFAOYSA-N
4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamine
10041227
4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1

It is an intermediate for the preparation of Elacridar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for the preparation of Elacridar.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 312.41
Percent Purity 97%
Chemical Name or Material 4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'