Learn More
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%
Product Code. p-7045127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11377466 1 g 1g
11387466 5 g 5g
2 options
Product Code. 11377466
Brand: Thermo Scientific Alfa Aesar L09516.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
CAS: 1547-36-0 | C5H4F6O3 | 226.07 g/mol
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1547-36-0
Molecular Formula C5H4F6O3
Molecular Weight (g/mol) 226.07
MDL Number MFCD00040910
InChI Key NOHJBOWARMTILE-UHFFFAOYSA-N
PubChem CID 15244
IUPAC Name 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
SMILES OC(=O)CC(O)(C(F)(F)F)C(F)(F)F
Specifications
Melting Point 85°C to 89°C
Quantity 1 g
UN Number UN3261
Beilstein 1911706
Synonym 3,3-bis trifluoromethyl-3-hydroxypropionic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butanoic acid, butyric acid, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethylbutyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, butanoic acid,4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, acmc-20anh2, 3,3-bis trifluoromethyl-3-hydroxypropanoic acid
Solubility Information Soluble in water.
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid