Learn More
4,4,4-Trifluorobutyric acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Melting Point | 29°C to 32°C |
Boiling Point | 166°C to 167°C |
Quantity | 5 g |
UN Number | UN3261 |
Solubility Information | Soluble in water. |
Formula Weight | 142.08 |
Percent Purity | 97% |
Chemical Name or Material | 4,4,4-Trifluorobutyric acid |