Learn More
4,4',4''-Trihydroxytriphenylmethane, 97%, Thermo Scientific™
Catalog Number p-4813035
Click to view available options
Quantity:
25 g
100 g
603-44-1
C19H16O3
292.33
MFCD00191589
WFCQTAXSWSWIHS-UHFFFAOYSA-N
4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
69047
4-[bis(4-hydroxyphenyl)methyl]phenol
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
603-44-1
C19H16O3
292.33
MFCD00191589
WFCQTAXSWSWIHS-UHFFFAOYSA-N
4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
69047
4-[bis(4-hydroxyphenyl)methyl]phenol
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Melting Point 244.0°C to 246.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 292.33
Percent Purity 97%
Chemical Name or Material 4, 4', 4''-Trihydroxytriphenylmethane
compliance-icons
Product Identifier
  • 4,4
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.