missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxychalcone, 99%

Product Code. p-7045405
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11332876

Brand: Thermo Scientific Alfa Aesar L10585.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2373-89-9 | C17H16O3 | 268.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2373-89-9
Molecular Formula C17H16O3
Molecular Weight (g/mol) 268.31
MDL Number MFCD00025815
InChI Key HDXVSZWKIHQDES-LFYBBSHMSA-N
Synonym 4,4'-dimethoxychalcone, e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 1,3-bis 4-methoxyphenyl prop-2-en-1-one, 4,4/'-dimethoxychalcone, 2-propen-1-one, 1,3-bis 4-methoxyphenyl, e-4,4'-dimethoxychalcone, 1,3-bis 4-methoxyphenyl-2-propen-1-one, e-1,3-bis 4-methoxyphenyl-2-propen-1-one, 2e-1,3-bis 4-methoxyphenyl-2-propen-1-one
PubChem CID 5377817
IUPAC Name (E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 99°C to 103°C
Quantity 10 g
Beilstein 1622293
Formula Weight 268.32
Percent Purity 99%
Chemical Name or Material 4,4'-Dimethoxychalcone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.