missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-1,3-cyclohexanedione, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 4,4-Dimethyl-1,3-cyclohexanedione |
| CAS | 562-46-9 |
| Molecular Formula | C8H12O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H12O2/c1-8(2)4-3-6(9)5-7(8)10/h3-5H2,1-2H3 |
| SMILES | CC1(C)CCC(=O)CC1=O |
| IUPAC Name | 4,4-dimethylcyclohexane-1,3-dione |
| Molecular Weight (g/mol) | 140.17 |
| Formula Weight | 140.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction