missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methyl-1-piperazinylmethyl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(4-Methyl-1-piperazinylmethyl)benzoic acid is used as Imatinib intermediate, as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, acids, bases.
4-(4-Methyl-1-piperazinylmethyl)benzoic acid is used as Imatinib intermediate, as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, acids, bases.
Specifications
Quantity | 250 mg |
Synonym | 4-4-methylpiperazin-1-ylmethyl benzoic acid, 4-4-methylpiperazin-1-yl methyl benzoic acid, 4-4-methyl-1-piperazinylmethyl benzoic acid, 4-4-methylpiperazinomethyl benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl methyl, benzoic acid,4-4-methyl-1-piperazinyl methyl, acmc-1bsqt, 4-4-methylpiperazinyl methyl benzoic acid, 4-4-methylpiperazino methyl benzoic acid |
Solubility Information | Slightly soluble in water. |
Formula Weight | 234.3 |
Percent Purity | 97% |
Chemical Name or Material | 4-(4-Methyl-1-piperazinylmethyl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'