missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-nitropyrimidine, TRC

Product Code. 30387920
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30387920 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30387920

Brand: Toronto Research Chemicals TRCD43564525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Dichloro-5-nitropyrimidine
CAS 4316-93-2
Molecular Formula C4 H Cl2 N3 O2
Quantity 25 g
InChI Formula InChI=1S/C4HCl2N3O2/c5-3-2(9(10)11)4(6)8-1-7-3/h1H
SMILES [O-][N+](=O)c1c(Cl)ncnc1Cl
IUPAC Name 4,6-dichloro-5-nitropyrimidine
Molecular Weight (g/mol) 193.98
Formula Weight 192.9446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.