missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine-5-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7055403
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
87600-98-4
C5H2Cl2N2O2
192.983
MFCD09909611
KGBLYFGHJNGQBK-UHFFFAOYSA-N
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro
24820487
4,6-dichloropyrimidine-5-carboxylic acid
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl
This item is not returnable. View return policy
87600-98-4
C5H2Cl2N2O2
192.983
MFCD09909611
KGBLYFGHJNGQBK-UHFFFAOYSA-N
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro
24820487
4,6-dichloropyrimidine-5-carboxylic acid
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl

4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 192.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,6-Dichloropyrimidine-5-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'