missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Difluoro-1,3-isobenzofurandione (Technical Grade, ~90%), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 4,7-Difluoro-1,3-isobenzofurandione (Technical Grade) |
| CAS | 652-40-4 |
| Molecular Formula | C8H2F2O3 |
| Quantity | 500 mg |
| Synonym | 3,6-Difluorophthalic Anhydride, 4,7-Difluoroisobenzofuran-1,3-dione, 3,6-Difluorophthalic Anhydride |
| InChI Formula | InChI=1S/C8H2F2O3/c9-3-1-2-4(10)6-5(3)7(11)13-8(6)12/h1-2H |
| SMILES | Fc1ccc(F)c2C(=O)OC(=O)c12 |
| IUPAC Name | 4,7-difluoro-2-benzofuran-1,3-dione |
| Molecular Weight (g/mol) | 184.1 |
| Formula Weight | 183.9972 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction