Learn More
Alfa Aesar™ 4-Amino-2-chloro-5-iodopyridine, 95%
Brand: Alfa Aesar™ H33926.MD
Additional Details : CAS : 800402-12-4 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
800402-12-4 | |
254.455 | |
DEJUUKULVAIMNF-UHFFFAOYSA-N | |
45480366 | |
C1=C(C(=CN=C1Cl)I)N |
C5H4ClIN2 | |
MFCD09834661 | |
2-chloro-5-iodo-4-pyridinamine, 4-amino-2-chloro-5-iodopyridine, 4-pyridinamine, 2-chloro-5-iodo, 2-chloro-5-iodo-4-aminopyridine, 2-chloro-5-iodo-pyridin-4-ylamine, 2-chloro-5-iodo-pyridine-4-ylamine, 2-chloro-5-iodo-4-pyridylamine, 2-chloro-4-amino-5-iodopyridine, 2-chloro-5-iodo-4-pyridineamine, 2-chloro-5-iodo-pyridin-4-amine | |
2-chloro-5-iodopyridin-4-amine |
Specifications
4-Amino-2-chloro-5-iodopyridine | |
126°C to 127°C | |
800402-12-4 | |
MFCD09834661 | |
DEJUUKULVAIMNF-UHFFFAOYSA-N | |
2-chloro-5-iodopyridin-4-amine | |
45480366 | |
95% |
Light sensitive | |
250mg | |
C5H4ClIN2 | |
2-chloro-5-iodo-4-pyridinamine, 4-amino-2-chloro-5-iodopyridine, 4-pyridinamine, 2-chloro-5-iodo, 2-chloro-5-iodo-4-aminopyridine, 2-chloro-5-iodo-pyridin-4-ylamine, 2-chloro-5-iodo-pyridine-4-ylamine, 2-chloro-5-iodo-4-pyridylamine, 2-chloro-4-amino-5-iodopyridine, 2-chloro-5-iodo-4-pyridineamine, 2-chloro-5-iodo-pyridin-4-amine | |
C1=C(C(=CN=C1Cl)I)N | |
254.455 | |
254.46 |