missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-mercaptopyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-2700536
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
333-49-3
C4H5N3S
127.17
DCPSTSVLRXOYGS-UHFFFAOYSA-N
2-thiocytosine, 4-amino-2-mercaptopyrimidine, thiocytosine, 4-aminopyrimidine-2-thiol, cytosine, thio, cytosine, 2-thio, 2 1h-pyrimidinethione, 4-amino, 2 1h-pyrimidinone, 4-aminothio, 4-amino-2 1h-pyrimidinethione, 4-amino-2-thiopyrimidine
2724245
6-amino-1H-pyrimidine-2-thione
C1=C(NC(=S)N=C1)N
This item is not returnable. View return policy
333-49-3
C4H5N3S
127.17
DCPSTSVLRXOYGS-UHFFFAOYSA-N
2-thiocytosine, 4-amino-2-mercaptopyrimidine, thiocytosine, 4-aminopyrimidine-2-thiol, cytosine, thio, cytosine, 2-thio, 2 1h-pyrimidinethione, 4-amino, 2 1h-pyrimidinone, 4-aminothio, 4-amino-2 1h-pyrimidinethione, 4-amino-2-thiopyrimidine
2724245
6-amino-1H-pyrimidine-2-thione
C1=C(NC(=S)N=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 127.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-mercaptopyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'