missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%
Brand: Alfa Aesar™ H62654.MD
Additional Details : Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
939968-08-8 | |
255.072 | |
JGOZEXIYNJERIP-UHFFFAOYSA-N | |
59231931 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)N)F |
C12H16BF2NO2 | |
MFCD18434474 | |
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3,5-difluorophenyl boronic acid pinacol ester, 4-amino-3,5-difluorobenzeneboronic acid pinacol ester, 4-amino-3,5-difluorophenylboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxabor, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester | |
250mg | |
C12H16BF2NO2 | |
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3,5-difluorophenyl boronic acid pinacol ester, 4-amino-3,5-difluorobenzeneboronic acid pinacol ester, 4-amino-3,5-difluorophenylboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxabor, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)N)F | |
255.072 | |
255.07 | |
96% |
White | |
939968-08-8 | |
MFCD18434474 | |
JGOZEXIYNJERIP-UHFFFAOYSA-N | |
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
59231931 | |
Solid |