missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-nitrodiphenyl sulfide, Thermo Scientific™

Product Code. p-7238579 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17565525

Brand: Thermo Scientific Acros 104180010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101-59-7
Molecular Formula C12H10N2O2S
Molecular Weight (g/mol) 246.28
MDL Number MFCD00007881
InChI Key ZBPKGHOGUVVDLF-UHFFFAOYSA-N
IUPAC Name 4-[(4-nitrophenyl)sulfanyl]aniline
SMILES NC1=CC=C(SC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Alpha Vector AMINONITRODIPHENYLSULFIDE
Melting Point 141°C to 145°C
Color Light yellow to brown
Infrared Spectrum Conforms
Assay Percent Range ≥97.5% (Titration with HClO4)
Packaging Glass Bottle
Linear Formula H2NC6H4SC6H4NO2
Quantity 1 g
Beilstein 13-534
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4'-nitrodiphenyl sulfide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.