missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoazobenzene-4'-sulfonic acid sodium salt, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7058380
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2491-71-6
C12H10N3NaO3S
299.28
MFCD00035564
FIXVWFINKCQNFG-UHFFFAOYSA-M
yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8
23691997
sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate
C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
2491-71-6
C12H10N3NaO3S
299.28
MFCD00035564
FIXVWFINKCQNFG-UHFFFAOYSA-M
yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8
23691997
sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate
C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Beilstein 8098548
Sensitivity Very Hygroscopic
Formula Weight 299.29
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Aminoazobenzene-4'-sulfonic acid sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'