missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-63775
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
14548-46-0
C12H9NO
183.21
MFCD00006430
SKFLCXNDKRUHTA-UHFFFAOYSA-N
4-benzoylpyridine, phenyl pyridin-4-yl methanone, phenyl 4-pyridyl ketone, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, 4-pyridyl phenyl ketone, pyridine, 4-benzoyl, ketone, phenyl 4-pyridyl, gamma-benzoylpyridine, 4-benzoyl pyridine
26731
phenyl(pyridin-4-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
This item is not returnable. View return policy
14548-46-0
C12H9NO
183.21
MFCD00006430
SKFLCXNDKRUHTA-UHFFFAOYSA-N
4-benzoylpyridine, phenyl pyridin-4-yl methanone, phenyl 4-pyridyl ketone, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, 4-pyridyl phenyl ketone, pyridine, 4-benzoyl, ketone, phenyl 4-pyridyl, gamma-benzoylpyridine, 4-benzoyl pyridine
26731
phenyl(pyridin-4-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 68.0°C to 72.0°C
Color Yellow
Boiling Point 315.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO(C5H4N)
Quantity 5 g
Beilstein 21, 331
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,very soluble in ether & ethanol
Formula Weight 183.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Benzoylpyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzoylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'