missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11446506
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
53937-02-3
C12H11NO2
201.23
MFCD04039771
DOVNUEPFPBWTSV-UHFFFAOYSA-N
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one
3607724
O=C1NC=CC(OCC2=CC=CC=C2)=C1
This item is not returnable. View return policy
53937-02-3
C12H11NO2
201.23
MFCD04039771
DOVNUEPFPBWTSV-UHFFFAOYSA-N
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one
3607724
O=C1NC=CC(OCC2=CC=CC=C2)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 194°C to 198°C
Quantity 1 g
IUPAC Name 4-(benzyloxy)-1,2-dihydropyridin-2-one
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material 4-Benzyloxy-2-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'