missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzaldehyde, TRC

Product Code. 30344023
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344023 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344023 Supplier Toronto Research Chemicals Supplier No. TRCB285876250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzyloxybenzaldehyde
CAS 4397-53-9
Molecular Formula C14H12O2
Quantity 250 g
Synonym 4-(Benzyloxy)benzaldehyde, 4-(Phenylmethoxy)benzaldehyde, NSC 131669, NSC 28298, p-(Benzyloxy)benzaldehyde
InChI Formula InChI=1S/C14H12O2/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-10H,11H2
SMILES O=Cc1ccc(OCc2ccccc2)cc1
IUPAC Name 4-phenylmethoxybenzaldehyde
Molecular Weight (g/mol) 212.24
Formula Weight 212.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.