missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-fluoro-2-nitrobenzene, 98%

Product Code. 11400727
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11400727

Brand: Thermo Scientific Alfa Aesar B22523.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 364-73-8 | C6H3BrFNO2 | 220.00 g/mol

4-Bromo-1-fluoro-2-nitrobenzene is used in the synthesis of anti-inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1-fluoro-2-nitrobenzene is used in the synthesis of anti-inflammatory agents.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-73-8
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD00129165
InChI Key UQEANKGXXSENNF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoronitrobenzene, 1-bromo-4-fluoro-3-nitrobenzene, 2-fluoro-5-bromonitrobenzene, 4-bromo-1-fluoro-2-nitro-benzene, benzene, 4-bromo-1-fluoro-2-nitro, pubchem1046, 5-bromo-fluoronitrobenzene, acmc-1cpc6, 5-bromo-2-fluronitrobenzene, ksc226i7f
PubChem CID 2736328
IUPAC Name 4-bromo-1-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Br)=CC=C1F

Specifications

Melting Point 18°C to 19°C
Density 1.786
Boiling Point 240°C to 241°C
Flash Point >110°C (230°F)
Refractive Index 1.5768
Quantity 25 g
UN Number UN2810
Beilstein 2329084
Solubility Information Difficult to mix.
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.