missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7058179
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
885702-34-1
C4H2BrClN2
193.43
MFCD08272196
FBEBVAQOMVWORE-UHFFFAOYSA-N
pyrimidine, 4-bromo-2-chloro-9ci, pyrimidine, 4-bromo-2-chloro, 2-chloro-4-bromopyrimidine
45117394
4-bromo-2-chloropyrimidine
ClC1=NC=CC(Br)=N1
This item is not returnable. View return policy
885702-34-1
C4H2BrClN2
193.43
MFCD08272196
FBEBVAQOMVWORE-UHFFFAOYSA-N
pyrimidine, 4-bromo-2-chloro-9ci, pyrimidine, 4-bromo-2-chloro, 2-chloro-4-bromopyrimidine
45117394
4-bromo-2-chloropyrimidine
ClC1=NC=CC(Br)=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an primary and secondary intermediate for pharmaceutical, organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 10334333
Solubility Information Slightly soluble in water.
Formula Weight 193.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chloropyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'