missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. p-7028258
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11407156

Brand: Thermo Scientific Alfa Aesar B25475.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153556-42-4
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00672932
InChI Key RMYOGXPGIDWJLU-UHFFFAOYSA-N
Synonym 3-fluoro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-fluoro, 3-fluoro-4-bromo benzoic acid, 4-bromo-3-fluoro-benzoic acid, pubchem1316, intermediates-zcf02169, acmc-1c1h6, ksc497g7p, 4-bromo-3-fluoro benzoic acid
PubChem CID 2773341
IUPAC Name 4-bromo-3-fluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)Br

Specifications

Melting Point 210°C to 213°C
Quantity 1 g
Formula Weight 219.01
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3-fluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.