missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-(trifluoromethyl)acetanilide, 97%

Product Code. 11408797
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
10 g
missing translation for 'unitSize'
10g
2.5g
Product Code. Quantity unitSize
11408797 2.5 g 2.5g
11418797 10 g 10g
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 11408797

missing translation for 'mfr': Thermo Scientific Alfa Aesar A19939.88

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 41513-05-7 | C9H7BrF3NO | 282.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41513-05-7
Molecular Formula C9H7BrF3NO
Molecular Weight (g/mol) 282.06
MDL Number MFCD00052361
InChI Key AOHSBSXIGFIMNC-UHFFFAOYSA-N
Synonym n-4-bromo-3-trifluoromethyl phenyl acetamide, 5-acetamido-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl acetanilide, 4'-bromo-3'-trifluoromethyl acetanilide, n-4-bromo-3-trifluoromethyl phenyl acet amide, n1-4-bromo-3-trifluoromethyl phenyl acetamide, pubchem2757, maybridge4_003018, buttpark 37\11-74
PubChem CID 697838
IUPAC Name N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)C(F)(F)F

Specifications

Melting Point 116°C to 119°C
Quantity 2.5 g
Formula Weight 282.07
Percent Purity 97%
Chemical Name or Material 4'-Bromo-3'-(trifluoromethyl)acetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.