missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62840.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407906

  • 487.00€ / 1g


Estimated Shipment
17-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

1150271-74-1
314.989
2-4-bromomethyl-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-3-fluorophenylboronic acid, pinacol ester, 4-bromomethyl-3-fluorobenzeneboronic acid, pinacol ester, 4-bromomethyl-3-fluorophenyl boronic acid pinacol ester
2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C13H17BBrFO2
OOTPRFIHAFABSN-UHFFFAOYSA-N
46739422
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CBr)F

Specifications

4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester
1150271-74-1
2-4-bromomethyl-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-3-fluorophenylboronic acid, pinacol ester, 4-bromomethyl-3-fluorobenzeneboronic acid, pinacol ester, 4-bromomethyl-3-fluorophenyl boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CBr)F
314.989
314.99
96%
1g
C13H17BBrFO2
OOTPRFIHAFABSN-UHFFFAOYSA-N
2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46739422
Solid
Videos
SDS Documents