missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl methyl sulfone, 99%, ACROS Organics™

Brand:  Acros Organics 355130250

Additional Details : CAS : 3466-32-8 Poids : 0.07500kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10599643

  • 189.00€ / 25g


Expédition estimée
10-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

3466-32-8
235.1
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
77014
CS(=O)(=O)C1=CC=C(C=C1)Br
C7H7BrO2S
MFCD00025065
4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
1-bromo-4-methylsulfonylbenzene

Specifications

4-Bromophenyl methyl sulfone
Glass bottle
102°C to 105°C
3466-32-8
100.0
C7H7BrO2S
4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
CS(=O)(=O)C1=CC=C(C=C1)Br
235.1
235.1
99%
Authentic
White
25g
98.5
98.5% min. (GC)
MFCD00025065
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
1-bromo-4-methylsulfonylbenzene
77014
Crystals or Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromophenyl methyl sulfone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents