Learn More
Applied Biosystems™ 3130/3100-Avant Genetic Analyzer 4-Capillary Array, 80 cm
Shop All Applied Biosystems Products
Click to view available options
Unit Size:
Each
Description
This is a 4-capillary array (internally uncoated) for the 3130 and 3100 Avant™ Genetic Analyzer.
Order Info
Each
Specifications
Specifications
| Quantity | 1 array |
| Content And Storage | Store the array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide. We guarantee the performance of the 3130 Capillary arrays up to 100 runs and within 1 year of purchase. |
| Capillary Length | 80 cm |
| Product Line | Avant |
| Shipping Condition | Room Temperature |
| Run Time | 170 min. |
| For Use With (Equipment) | 3100-Avant Genetic Analyzer, 3130 Genetic Analyzer |
| For Use With (Application) | Sequencing, Fragment Analysis |