missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2-(4-morpholinyl)quinazoline, 97%

Brand:  Alfa Aesar™ H63458.03

Additional Details : CAS : 39216-67-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410169

  • 303.00€ / 1g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

39216-67-6
249.698
4-chloro-2-morpholinoquinazoline, 4-chloro-2-morpholin-4-yl quinazoline, 4-4-chloroquinazolin-2-yl morpholine, 4-chloro-2-4-morpholinyl quinazoline
4-(4-chloroquinazolin-2-yl)morpholine
C12H12ClN3O
UWAUCVLFWGAOGM-UHFFFAOYSA-N
12286589
C1COCCN1C2=NC3=CC=CC=C3C(=N2)Cl

Specifications

4-Chloro-2-(4-morpholinyl)quinazoline
39216-67-6
4-chloro-2-morpholinoquinazoline, 4-chloro-2-morpholin-4-yl quinazoline, 4-4-chloroquinazolin-2-yl morpholine, 4-chloro-2-4-morpholinyl quinazoline
C1COCCN1C2=NC3=CC=CC=C3C(=N2)Cl
249.698
249.7
97%
1g
C12H12ClN3O
UWAUCVLFWGAOGM-UHFFFAOYSA-N
4-(4-chloroquinazolin-2-yl)morpholine
12286589
Solid
Videos
SDS Documents