missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine, 96%, Thermo Scientific™

Catalog Number p-7032840
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
610274-01-6
C12H13ClN2S
252.76
MFCD03085777
DNHPYCAMLHXYLU-UHFFFAOYSA-N
4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-benzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-1 benzothiolo 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine, 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6-tetraene, 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene
3860934
4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-[1]benzothiolo[2,3-d]pyrimidine
CC1CCC2=C(C1)SC3=C2C(=NC(=N3)C)Cl
This item is not returnable. View return policy
610274-01-6
C12H13ClN2S
252.76
MFCD03085777
DNHPYCAMLHXYLU-UHFFFAOYSA-N
4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-benzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-1 benzothiolo 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine, 4-chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine, 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6-tetraene, 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene
3860934
4-chloro-2,7-dimethyl-5,6,7,8-tetrahydro-[1]benzothiolo[2,3-d]pyrimidine
CC1CCC2=C(C1)SC3=C2C(=NC(=N3)C)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 91°C to 95°C
Quantity 250 mg
Formula Weight 252.77
Percent Purity 96%
Chemical Name or Material 4-Chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'