missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzeneboronic acid, 98%

Product Code. 15449267
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 15449267

missing translation for 'mfr': Thermo Scientific Alfa Aesar H52529.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 762287-57-0 | C7H8BClO3 | 186.398 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762287-57-0
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.398
MDL Number MFCD02684317
InChI Key NZRRMTBNTSBIFH-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxyphenyl boronic acid, 4-chloro-2-methoxybenzeneboronic acid, 4-chloro-2-methoxy-phenyl boronic acid, boronic acid, 4-chloro-2-methoxyphenyl, acmc-209p2g, ksc495a8j, ablock ab-12-6332, 2-methoxy-4-chlorophenylboronic acid, 4-chloranyl-2-methoxy-phenyl boronic acid, 4-chloro-2-methyloxy phenyl boronic acid
PubChem CID 3858606
IUPAC Name (4-chloro-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)OC)(O)O

Specifications

Melting Point 128°C to 134°C
Quantity 250 mg
Formula Weight 186.4
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methoxybenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.