missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 11372136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372136 5 g 5g
11362136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372136 Supplier Thermo Scientific Alfa Aesar Supplier No. L03413.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-3,5-dinitrobenzonitrile is may be used in the synthesis of 3-nitro-4-thiocyanobenzonitryl. It is also used as Pharmaceutical intermediates .

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong bases.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1930-72-9
Molecular Formula C7H2ClN3O4
Molecular Weight (g/mol) 227.56
MDL Number MFCD00007067
InChI Key SCGDEDHSPCXGEC-UHFFFAOYSA-N
Synonym benzonitrile, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzenenitrile, 1-chloro-4-cyano-2,6-dinitrobenzene, 4-chloro-3,5-dinitrobenzenecarbonitrile, acmc-1brcu, 4-chloro-3,5-dinitrobenzonitril, 3,5-dinitro-4-chlorobenzonitrile, 4-cyano-2,6-dinitrochlorobenzene, 4-chloro-3,5-dinitro-benzonitrile, benzonitrile,4-chloro-3,5-dinitro
PubChem CID 16008
IUPAC Name 4-chloro-3,5-dinitrobenzonitrile
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C#N

Specifications

Melting Point 140°C to 142°C
Quantity 5 g
UN Number UN3439
Beilstein 1990451
Solubility Information Insoluble in water.
Formula Weight 227.56
Percent Purity 97%
Chemical Name or Material 4-Chloro-3,5-dinitrobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.