missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(cyclopropylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15409905
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871332-73-9
C10H11BClNO3
239.462
MFCD07783865
AZBIVGURALLESY-UHFFFAOYSA-N
4-chloro-3-cyclopropylcarbamoyl phenylboronic acid, 4-chloro-3-cyclopropylcarbamoyl phenyl boronic acid, acmc-209qh3, n-cyclopropyl 5-borono-2-chlorobenzamide, n-cyclopropyl 2-chloro-5-boronobenzamide, 4-chloro-3-cyclopropylcarbamoyl benzeneboronic acid, boronic acid,b-4-chloro-3-cyclopropylamino carbonyl phenyl
44886936
[4-chloro-3-(cyclopropylcarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)Cl)C(=O)NC2CC2)(O)O
This item is not returnable. View return policy
871332-73-9
C10H11BClNO3
239.462
MFCD07783865
AZBIVGURALLESY-UHFFFAOYSA-N
4-chloro-3-cyclopropylcarbamoyl phenylboronic acid, 4-chloro-3-cyclopropylcarbamoyl phenyl boronic acid, acmc-209qh3, n-cyclopropyl 5-borono-2-chlorobenzamide, n-cyclopropyl 2-chloro-5-boronobenzamide, 4-chloro-3-cyclopropylcarbamoyl benzeneboronic acid, boronic acid,b-4-chloro-3-cyclopropylamino carbonyl phenyl
44886936
[4-chloro-3-(cyclopropylcarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)Cl)C(=O)NC2CC2)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 238°C to 242°C
Quantity 1 g
Formula Weight 239.46
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-(cyclopropylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'