missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,4-dichlorophenyl)butyramide, 97%

Brand:  Alfa Aesar™ H59853.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406288

  • 522.00€ / 1g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

457618-46-1
266.546
SPTDUZNBZNYZFE-UHFFFAOYSA-N
4396570
C1=CC(=C(C=C1Cl)Cl)NC(=O)CCCCl
C10H10Cl3NO
MFCD03361724
4-chloro-n-2,4-dichlorophenyl butanamide, 4-chloro-n-2,4-dichlorophenyl butyramide
4-chloro-N-(2,4-dichlorophenyl)butanamide

Specifications

4-Chloro-N-(2,4-dichlorophenyl)butyramide
457618-46-1
MFCD03361724
SPTDUZNBZNYZFE-UHFFFAOYSA-N
4-chloro-N-(2,4-dichlorophenyl)butanamide
4396570
97%
1g
C10H10Cl3NO
4-chloro-n-2,4-dichlorophenyl butanamide, 4-chloro-n-2,4-dichlorophenyl butyramide
C1=CC(=C(C=C1Cl)Cl)NC(=O)CCCCl
266.546
266.55
Videos
SDS Documents