missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15411256
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1221823-27-3
C13H20BNO4S
297.176
MFCD20265279
JDFPAMLPRYKLRV-UHFFFAOYSA-N
4-cyclohexylmethylsulfamoyl benzeneboronic acid, 4-cyclohexylmethylsulfamoyl phenylboronic acid
57655465
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O
This item is not returnable. View return policy
1221823-27-3
C13H20BNO4S
297.176
MFCD20265279
JDFPAMLPRYKLRV-UHFFFAOYSA-N
4-cyclohexylmethylsulfamoyl benzeneboronic acid, 4-cyclohexylmethylsulfamoyl phenylboronic acid
57655465
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 297.18
Percent Purity 97%
Chemical Name or Material 4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'