Learn More
4-(Diazonium)benzenesulfonic Acid, Fluoroborate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Diazonium)benzenesulfonic Acid, Fluoroborate Salt |
| CAS | 2145-24-6 |
| Molecular Formula | C6 H5 N2 O3 S . B F4 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H4N2O3S.BF4/c7-8-5-1-3-6(4-2-5)12(9,10)11;2-1(3,4)5/h1-4H;/q;-1/p+1 |
| SMILES | OS(=O)(=O)c1ccc(cc1)[N+]#N.F[B-](F)(F)F |
| IUPAC Name | 4-sulfobenzenediazonium;tetrafluoroborate |
| Molecular Weight (g/mol) | 271.99 |
| Formula Weight | 272.005 |