missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Dimethylamino)pyridine tribromide, 97%

Brand:  Alfa Aesar™ H55010.22

Additional Details : CAS : 92976-81-3 Weight : 0.10000kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 15411128

  • 329.00€ / 100g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

92976-81-3
361.883
TYDZHQMKSLAFKM-UHFFFAOYSA-N
131863682
C7H10Br3N2-
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br

Specifications

4-(Dimethylamino)pyridine tribromide
100g
C7H10Br3N2-
UN3261
TYDZHQMKSLAFKM-UHFFFAOYSA-N
361.883
362.89
129°C to 132°C
92976-81-3
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br
131863682
97%
Videos
SDS Documents