missing translation for 'onlineSavingsMsg'
Learn More

4-epi-Tetracycline Hydrochloride, TRC

Product Code. 30425346
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30425346 5 mg 5mg
1 options
This item is not returnable. View return policy

Product Code. 30425346

Brand: Toronto Research Chemicals TRCT2914055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Epitetracycline hydrochloride
CAS 23313-80-6
Purity Grade Notes HPLC
Molecular Formula C22 H24 N2 O8 . Cl H
Quantity 5 mg
Synonym 4-Epitetracycline hydrochloride, Chlortetracycline Hydrochloride Imp. D (EP) as Hydrochloride, (4R,4aS,5aS,6S,12aS)-4-(Dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride, 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','hydrochloride (1:1)','(4R,4aS,5aS,6S,12aS)-, 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','(4R,4aS,5aS,6S,12aS)- (9CI), 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','4-epimer (8CI), 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','[4R-(4α,4aβ,5aβ,6α,12aβ)]-, 4-Epitetracycline monohydrochloride, 4-epi-Tetracycline hydrochloride, Epitetracycline hydrochloride
InChI Formula InChI=1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15+,21+,22-;/m0./s1
SMILES Cl.CN(C)[C@@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O
IUPAC Name (4R,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 480.9
Formula Weight 480.1299
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.