missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-epi-Tetracycline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Epitetracycline hydrochloride |
| CAS | 23313-80-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H24 N2 O8 . Cl H |
| Quantity | 50 mg |
| Synonym | 4-Epitetracycline hydrochloride, Chlortetracycline Hydrochloride Imp. D (EP) as Hydrochloride, (4R,4aS,5aS,6S,12aS)-4-(Dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride, 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','hydrochloride (1:1)','(4R,4aS,5aS,6S,12aS)-, 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','(4R,4aS,5aS,6S,12aS)- (9CI), 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','4-epimer (8CI), 2-Naphthacenecarboxamide','4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-','monohydrochloride','[4R-(4α,4aβ,5aβ,6α,12aβ)]-, 4-Epitetracycline monohydrochloride, 4-epi-Tetracycline hydrochloride, Epitetracycline hydrochloride |
| InChI Formula | InChI=1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15+,21+,22-;/m0./s1 |
| SMILES | Cl.CN(C)[C@@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O |
| IUPAC Name | (4R,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 480.9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?