missing translation for 'onlineSavingsMsg'
Learn More

4-Epitetracycline Hydrochloride, Mikromol™

Product Code. 17943096
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
17943096 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 17943096

Brand: Mikromol MM0452.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Epitetracycline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Epitetracycline hydrochloride
CAS 23313-80-6
Analyte or Component Names 4-Epitetracycline hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H24 N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Chlortetracycline Hydrochloride Imp. D (EP) as Hydrochloride, (4R,4aS,5aS,6S,12aS)-4-(Dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide Hydrochloride, 4-Epitetracycline Hydrochloride, Chlortetracycline Hydrochloride Impurity D as Hydrochloride, Tetracycline Hydrochloride Impurity A as Hydrochloride, Chlortetracycline Impurity D as Hydrochloride, Tetracycline Impurity A as Hydrochloride
InChI Formula InChI=1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15+,21+,22-;/m0./s1
SMILES Cl.CN(C)[C@@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O
IUPAC Name (4R,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 480.9
Quantity 50 mg
Formula Weight 480.1299 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.