missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7052481
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1373920-79-6
C9H7ClF2O2
220.6
MFCD22201053
DFCDPQTYYGJYOD-UHFFFAOYSA-N
86277653
4-ethoxy-2,6-difluorobenzoyl chloride
CCOC1=CC(=C(C(=C1)F)C(=O)Cl)F
This item is not returnable. View return policy
1373920-79-6
C9H7ClF2O2
220.6
MFCD22201053
DFCDPQTYYGJYOD-UHFFFAOYSA-N
86277653
4-ethoxy-2,6-difluorobenzoyl chloride
CCOC1=CC(=C(C(=C1)F)C(=O)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'