missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isothiocyanate 97%, Thermo Scientific™

Catalog Number p-4391333
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
3460-49-9
C9H9NOS
179.237
REEKOMRZYJXXNR-UHFFFAOYSA-N
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
77006
1-ethoxy-4-isothiocyanatobenzene
CCOC1=CC=C(C=C1)N=C=S
This item is not returnable. View return policy
3460-49-9
C9H9NOS
179.237
REEKOMRZYJXXNR-UHFFFAOYSA-N
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
77006
1-ethoxy-4-isothiocyanatobenzene
CCOC1=CC=C(C=C1)N=C=S
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 4-Ethoxyphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'