missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-trans-beta-styrylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15497775
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15497775

Brand: Thermo Scientific Alfa Aesar H30744.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870717-91-2
Molecular Formula C16H23BO2
Molecular Weight (g/mol) 258.17
MDL Number MFCD08276837
InChI Key WXXFAINZQODXKD-UHFFFAOYSA-N
Synonym e-2-4-ethylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-2-4-ethylphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-2-4-ethylphenyl vinylboronic acid pinacol ester, trans-2-4-ethylphenyl vinylboronic acid pinacol ester, 2-e-2-4-ethylphenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-2-4-ethylphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole
PubChem CID 16218145
IUPAC Name 2-[(E)-2-(4-ethylphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1

Specifications

Melting Point 46°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 258.17
Percent Purity 97%
Chemical Name or Material 4-Ethyl-trans-beta-styrylboronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.