missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7034096
69.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1034287-04-1
C14H17BO2
228.098
MFCD16294504
LOVNTFMVZVIASV-UHFFFAOYSA-N
2-4-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-ethynylbenzeneboronic acid pinacol ester, amtb205, 4-ethynylphenylboronic acid pinacol ester
57415690
2-(4-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1034287-04-1
C14H17BO2
228.098
MFCD16294504
LOVNTFMVZVIASV-UHFFFAOYSA-N
2-4-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-ethynylbenzeneboronic acid pinacol ester, amtb205, 4-ethynylphenylboronic acid pinacol ester
57415690
2-(4-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 228.1
Percent Purity 95%
Chemical Name or Material 4-Ethynylbenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'