Learn More
Bochem™ 4-Finger Malleable Cast Iron Condenser Clamp
Used in the process of cooling hot vapors and liquid. BOCHEM™ 4-Finger Malleable Cast Iron Condenser Clamp offers secure immobilization of laboratory glassware.
Brand:  Bochem™ 5474
Additional Details : Weight : 0.35100kg
Product Code. 10768571
  • 37.35€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Malleable clast iron
  • Chromium plated
  • Swivel type
  • Silicone coated
Specifications
Specifications
1 Ea.
Malleable Cast Iron
150 mm
Condenser Clamp
50 to 100 mm
50 to 100 mm