missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 11485736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11485736 5 g 5g
11475736 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11485736

Brand: Thermo Scientific Alfa Aesar B20814.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67515-61-1
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.13
MDL Number MFCD00061171
InChI Key DGBDVJVPBJMVDH-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl benzyl alcohol, 4-fluoro-3-trifluoromethyl phenyl methanol, 3-trifluoromethyl-4-fluorobenzyl alcohol, 4-fluoro-3-trifluoromethyl phenyl methan-1-ol, rarechem al bd 0323, 4-fluoro-3-trifluoromethyl benzylalcohol, benzenemethanol,4-fluoro-3-trifluoromethyl, 4-fluoranyl-3-trifluoromethyl phenyl methanol
PubChem CID 144256
IUPAC Name [4-fluoro-3-(trifluoromethyl)phenyl]methanol
SMILES OCC1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Boiling Point 80°C to 85°C (0.1 mmHg)
Refractive Index 1.45
Quantity 5 g
Beilstein 1960409
Formula Weight 194.13
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.