Learn More
4'-Fluorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog Number p-4813118
Click to view available options
Quantity:
1 g
5 g
116748-66-4
C12H8ClFO2S
270.71
CIDMHDJTWVMBIF-UHFFFAOYSA-N
4'-fluorobiphenyl-4-sulfonyl chloride, 4-4-fluorophenyl benzenesulfonyl chloride, 4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride, 4'-fluoro-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro, 4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride, acmc-20apun, 4'-fluorobiphenyl-4-sulfonylchloride, 4'-fluoro-4-biphenylsulfonyl chloride
2794749
4-(4-fluorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F
116748-66-4
C12H8ClFO2S
270.71
CIDMHDJTWVMBIF-UHFFFAOYSA-N
4'-fluorobiphenyl-4-sulfonyl chloride, 4-4-fluorophenyl benzenesulfonyl chloride, 4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride, 4'-fluoro-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro, 4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride, acmc-20apun, 4'-fluorobiphenyl-4-sulfonylchloride, 4'-fluoro-4-biphenylsulfonyl chloride
2794749
4-(4-fluorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 82.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 270.71
Percent Purity 97%
Chemical Name or Material 4'-Fluorobiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 4'-Fluorobiphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.