missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-4813120
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
24589-77-3
C7H9ClN2O2
188.61
MFCD00039073
XHLQMKQBCHYRLC-UHFFFAOYSA-N
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride
90547
4-hydrazinylbenzoic acid;hydrochloride
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O
This item is not returnable. View return policy
24589-77-3
C7H9ClN2O2
188.61
MFCD00039073
XHLQMKQBCHYRLC-UHFFFAOYSA-N
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride
90547
4-hydrazinylbenzoic acid;hydrochloride
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 252.0°C to 254.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 188.61
Percent Purity 97%
Chemical Name or Material 4-Hydrazinobenzoic acid hydrochloride
compliance-icons
Product Identifier
  • 4-Hydrazinobenzoic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'