missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylthio-6-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™

Catalog Number 15410636
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
16097-62-4
C6H5F3N2OS
210.174
MFCD00153191
BRLPDRZKOMZVOY-UHFFFAOYSA-N
2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
248062
2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
CSC1=NC(=O)C=C(N1)C(F)(F)F
This item is not returnable. View return policy
16097-62-4
C6H5F3N2OS
210.174
MFCD00153191
BRLPDRZKOMZVOY-UHFFFAOYSA-N
2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
248062
2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
CSC1=NC(=O)C=C(N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 176°C to 180°C
Quantity 1 g
Formula Weight 210.17
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-2-methylthio-6-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'